N-(1,3-Benzodioxol-5-yl)-2-chloroacetamide 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(Benzo[d][1,3]dioxol-5-yl)-2-chloroacetamide
OR2919-500MG
APOSOR2919-500MGEA
21.9
EUR
APOSOR2919-500MG
N-(1,3-Benzodioxol-5-yl)-2-chloroacetamide 97%
N-(1,3-Benzodioxol-5-yl)-2-chloroacetamide
Formel:
C₉H₈ClNO₃ Molecular Weight: 213,62 g/mol Schmelzpunkt: 155…156 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00245737 CAS-Nummer: 227199-07-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...