2-Chlor-4-methoxybenzonitril 99%

Lieferant: Apollo Scientific
Achtung

OR29211-5G
APOSOR29211-5GEA 21.9 EUR
APOSOR29211-5G
2-Chlor-4-methoxybenzonitril 99%
2-Chlor-4-methoxybenzonitril
Formel: C₈H₆ClNO
Molecular Weight: 167,5900 g/mol
Schmelzpunkt: 75…77 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156133
CAS-Nummer: 127666-99-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR