2-Formylbenzene-1,4-disulphonate

Lieferant: Apollo Scientific
Achtung

OR2923-1G
APOSOR2923-1GEA 217 EUR
APOSOR2923-1G
2-Formylbenzene-1,4-disulphonate
2-Formylbenzene-1,4-disulphonate
Formel: C₇H₆O₇S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03840860
CAS-Nummer: 51818-11-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR