2-(1H-Pyrazol-3-yl)pyridin

Lieferant: Apollo Scientific
Achtung

OR29238-1G
APOSOR29238-1GEA 21.9 EUR
APOSOR29238-1G
2-(1H-Pyrazol-3-yl)pyridin
2-(1H-Pyrazol-3-yl)pyridin
Formel: C₈H₇N₃
Molecular Weight: 145,16 g/mol
Schmelzpunkt: 121…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00115151
CAS-Nummer: 75415-03-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR