ethyl 2-(1,4-dimethyl-3-oxo-2-piperazinyl)acetate

Lieferant: Apollo Scientific

OR29248-1G
APOSOR29248-1GEA 132 EUR
APOSOR29248-1G
ethyl 2-(1,4-dimethyl-3-oxo-2-piperazinyl)acetate
ethyl 2-(1,4-dimethyl-3-oxo-2-piperazinyl)acetate
Formel: C₁₀H₁₈N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00178772
CAS-Nummer: 175205-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR