3-Chloro-4-hydroxybenzoic acid hemihydrate

Lieferant: Apollo Scientific
Achtung

OR2925-100G OR2925-500G OR2925-50G
APOSOR2925-100GEA 271 EUR
APOSOR2925-100G APOSOR2925-500G APOSOR2925-50G
3-Chloro-4-hydroxybenzoic acid hemihydrate
3-Chloro-4-hydroxybenzoic acid hemihydrate
Formel: 2C₇H₅ClO₃·H₂O
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02662353
CAS-Nummer: 128161-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR