4-Pyrrol-1-ylphenylamine
Lieferant: Apollo Scientific
|
Achtung
|
OR29302-1G
OR29302-500MG
OR29302-5G
APOSOR29302-1GEA
29
EUR
APOSOR29302-1G
APOSOR29302-500MG
APOSOR29302-5G
4-Pyrrol-1-ylphenylamine
4-Pyrrol-1-ylphenylamine
Formel:
C₁₀H₁₀N₂ Schmelzpunkt: 80,7…81.2 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01934575 CAS-Nummer: 52768-17-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...