3,5-Dimethyl-p-anissäure 98%
Lieferant: Apollo Scientific
Synonyme:
3,5-Dimethyl-4-methoxybenzoesäure
OR29323-25G
APOSOR29323-25GEA
108
EUR
APOSOR29323-25G
3,5-Dimethyl-p-anissäure 98%
3,5-Dimethyl-p-anissäure
Formel:
C₁₀H₁₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00020309 CAS-Nummer: 21553-46-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...