4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Dioxo-4H,6H-pyrrolo[1,2-a][4,1]benzoxazepine, 4H,6H-Benzo[e]pyrrolo[2,1-c][1,4]oxazepine-4,6-dione

OR29335-20MG
APOSOR29335-20MGEA 29 EUR
APOSOR29335-20MG
4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione
4H,6H-Pyrrolo[1,2-a][4,1]benzoxazepine-4,6-dione
Formel: C₁₂H₇NO₃
Molecular Weight: 213,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00662464
CAS-Nummer: 259269-45-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR