2-(4-Benzylpiperazin-1-yl)benzaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR29343-1G
APOSOR29343-1GEA
92.8
EUR
APOSOR29343-1G
2-(4-Benzylpiperazin-1-yl)benzaldehyde
2-(4-Benzylpiperazin-1-yl)benzaldehyde
Formel:
C₁₈H₂₀N₂O Schmelzpunkt: 57…59 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
112253-26-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...