4-Phenylimidazol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Phenyl-1H-imidazole

OR29380-25G
APOSOR29380-25GEA 98.6 EUR
APOSOR29380-25G
4-Phenylimidazol 98%
4-Phenylimidazol
Formel: C₉H₈N₂
Molecular Weight: 144,18 g/mol
Schmelzpunkt: 128…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005197
CAS-Nummer: 670-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR