2-(2-Chlorobenzylthio)ethylamine

Lieferant: Apollo Scientific
Gefahr

OR29429-1G OR29429-250MG
APOSOR29429-1GEA 198 EUR
APOSOR29429-1G APOSOR29429-250MG
2-(2-Chlorobenzylthio)ethylamine
2-(2-Chlorobenzylthio)ethylamine
Formel: C₉H₁₂ClNS
Molecular Weight: 201.712 g/mol
Siedepunkt: 160…165 °C/13mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD01764494
CAS-Nummer: 42416-23-9
UN: 2735
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR