Ethyl 2-acetyl-4-oxopentanoate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-ethanoyl-4-oxopentanoate, 3-(Ethoxycarbonyl)hexane-2,5-dione

OR29456-500MG
APOSOR29456-500MGEA 143 EUR
APOSOR29456-500MG
Ethyl 2-acetyl-4-oxopentanoate
Ethyl 2-acetyl-4-oxopentanoate
Formel: C₉H₁₄O₄
Molecular Weight: 186,205 g/mol
Dichte: 1,06 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00204229
CAS-Nummer: 41892-81-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR