3-Aminoisonicotinsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR29468-25G
OR29468-5G
APOSOR29468-25GEA
102
EUR
APOSOR29468-25G
APOSOR29468-5G
3-Aminoisonicotinsäure 98%
3-Aminoisonicotinsäure
Formel:
C₆H₆N₂O₂ Schmelzpunkt: 287…291 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137840 CAS-Nummer: 7579-20-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...