Ethyl 5-(4-aminophenyl)-2-methyl-3-furoate

Lieferant: Apollo Scientific
Achtung

OR29475-250MG
APOSOR29475-250MGEA 109 EUR
APOSOR29475-250MG
Ethyl 5-(4-aminophenyl)-2-methyl-3-furoate
Ethyl 5-(4-aminophenyl)-2-methyl-3-furoate
Formel: C₁₄H₁₅NO₃
Molecular Weight: 245,274 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00276981
CAS-Nummer: 175276-70-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR