3-Thiophen-2-yl-1H-pyrazole-4-carbaldehyde 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(Thiophen-2-yl)-1H-pyrazole-4-carbaldehyde
OR29524-1G
APOSOR29524-1GEA
166
EUR
APOSOR29524-1G
3-Thiophen-2-yl-1H-pyrazole-4-carbaldehyde 98%
3-Thiophen-2-yl-1H-pyrazole-4-carbaldehyde
Formel:
C₈H₆N₂OS Molecular Weight: 178,21 g/mol Schmelzpunkt: 63…66 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD02677688 CAS-Nummer: 26033-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...