5-Chlorobenzo[b]thiophene-3-carboxaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Chlorobenzo[b]thiophene-3-carbaldehyde, 5-Chloro-3-formylbenzo[b]thiophene, 5-Chloro-1-benzothiophene-3-carboxaldehyde

OR29527-100MG
APOSOR29527-100MGEA 34.8 EUR
APOSOR29527-100MG
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
5-Chlorobenzo[b]thiophene-3-carboxaldehyde
Formel: C₉H₅ClOS
Molecular Weight: 196,653 g/mol
Schmelzpunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02677690
CAS-Nummer: 16296-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR