3,5-Dichlorbenzylalkohol
Lieferant: Apollo Scientific
|
Achtung
|
OR29565-25G
OR29565-5G
OR29565-100G
APOSOR29565-25GEA
29
EUR
APOSOR29565-25G
APOSOR29565-5G
APOSOR29565-100G
3,5-Dichlorbenzylalkohol
3,5-Dichlorbenzylalkohol
Formel:
C₇H₆Cl₂O Schmelzpunkt: 76…82 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004634 CAS-Nummer: 60211-57-6 EINECS: 262-109-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...