2,4'-Dichloracetanilid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(4-Chlorophenyl)-2-chloroacetamide, 2-Chloro-N-(4-chlorophenyl)acetamide, 2,4′-Dichloroacetanilide
OR29570-1G
APOSOR29570-1GEA
21.9
EUR
APOSOR29570-1G
2,4'-Dichloracetanilid
2,4'-Dichloracetanilid
Formel:
C₈H₇Cl₂NO Molecular Weight: 204.0533 g/mol Schmelzpunkt: 169…172 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00018904 CAS-Nummer: 3289-75-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...