2,3-Dichlorthiobenzamid
Lieferant: Apollo Scientific
Synonyme:
2,3-Dichlorthiobenzoesäureamid
OR29605-1G
APOSOR29605-1GEA
132
EUR
APOSOR29605-1G
2,3-Dichlorthiobenzamid
2,3-Dichlorthiobenzamid
Formel:
C₇H₅Cl₂NS Molecular Weight: 206,09 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00221163 CAS-Nummer: 84863-83-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...