5-Methyl-1H-pyrazole-3-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR29613-1G
APOSOR29613-1GEA
50.9
EUR
APOSOR29613-1G
5-Methyl-1H-pyrazole-3-carbohydrazide
5-Methyl-1H-pyrazole-3-carbohydrazide
Formel:
C₅H₈N₄O Molecular Weight: 140,15 g/mol Schmelzpunkt: 158…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03423274 CAS-Nummer: 40535-14-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...