3-tert-Butyl-1H-pyrazole-5-carbohydrazide
Lieferant: Apollo Scientific
|
Achtung
|
OR29615-250MG
APOSOR29615-250MGEA
58
EUR
APOSOR29615-250MG
3-tert-Butyl-1H-pyrazole-5-carbohydrazide
3-tert-Butyl-1H-pyrazole-5-carbohydrazide
Formel:
C₈H₁₄N₄O Molecular Weight: 182,23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00619318 CAS-Nummer: 262292-02-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...