4'-(4-Bromphenyl)acetophenon 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR2964-25G
APOSOR2964-25GEA
0
EUR
APOSOR2964-25G
4'-(4-Bromphenyl)acetophenon 98%
4'-(4-Bromphenyl)acetophenon
Formel:
C₁₄H₁₁BrO Schmelzpunkt: 129…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00143242 CAS-Nummer: 5731-01-1 UN: 3077 ADR: 9+EHS,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...