1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Ethyl-3-methylpyrazole-5-carboxylic acid

OR29643-25G
APOSOR29643-25GEA 94.4 EUR
APOSOR29643-25G
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154.17 g/mol
Schmelzpunkt: 142…143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156147
CAS-Nummer: 50920-65-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR