5-Methoxy-1,3-benzothiazol-2-amine 98%

Lieferant: Apollo Scientific
Achtung

OR2965-25G
APOSOR2965-25GEA 973 EUR
APOSOR2965-25G
5-Methoxy-1,3-benzothiazol-2-amine 98%
5-Methoxy-1,3-benzothiazol-2-amine
Formel: C₈H₈N₂OS
Lagertemperatur: Kühlschrank
MDL: MFCD00464314
CAS-Nummer: 54346-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR