4-(Chloromethyl)-2-methyl-1,3-thiazole 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Chloromethyl)-2-methylthiazole

OR29650-1G
APOSOR29650-1GEA 203 EUR
APOSOR29650-1G
4-(Chloromethyl)-2-methyl-1,3-thiazole 95%
4-(Chloromethyl)-2-methyl-1,3-thiazole
Formel: C₅H₆ClNS
Lagertemperatur: Kühlschrank
MDL: MFCD00051642
CAS-Nummer: 39238-07-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR