Thiophene-3-acetic acid hydrazide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Thien-3-yl)acetohydrazide

OR29655-1G
APOSOR29655-1GEA 132 EUR
APOSOR29655-1G
Thiophene-3-acetic acid hydrazide
Thiophene-3-acetic acid hydrazide
Formel: C₆H₈N₂OS
Molecular Weight: 156,21 g/mol
Schmelzpunkt: 85…86 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052946
CAS-Nummer: 175276-94-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR