2-[4-(4,5-Dihydro-1,3-thiazol-2-yl)phenoxy]acetonitrile

Lieferant: Apollo Scientific
Gefahr

OR29659-250MG
APOSOR29659-250MGEA 63.8 EUR
APOSOR29659-250MG
2-[4-(4,5-Dihydro-1,3-thiazol-2-yl)phenoxy]acetonitrile
2-[4-(4,5-Dihydro-1,3-thiazol-2-yl)phenoxy]acetonitrile
Formel: C₁₁H₁₀N₂OS
Molecular Weight: 218,275 g/mol
Schmelzpunkt: 83…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052942
CAS-Nummer: 175276-95-6
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR