2-Methyl-1,3-thiazole-4-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Cyano-2-methyl-1,3-thiazole, 2-Methylthiazole-4-carbonitrile
OR29666-1G
OR29666-250MG
APOSOR29666-1GEA
74.1
EUR
APOSOR29666-1G
APOSOR29666-250MG
2-Methyl-1,3-thiazole-4-carbonitrile
2-Methyl-1,3-thiazole-4-carbonitrile
Formel:
C₅H₄N₂S Molecular Weight: 124.164 g/mol Schmelzpunkt: 58…61 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00068103 CAS-Nummer: 21917-76-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...