2-Methyl-1,3-thiazole-4-carbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Cyano-2-methyl-1,3-thiazole, 2-Methylthiazole-4-carbonitrile

OR29666-1G OR29666-250MG
APOSOR29666-1GEA 74.1 EUR
APOSOR29666-1G APOSOR29666-250MG
2-Methyl-1,3-thiazole-4-carbonitrile
2-Methyl-1,3-thiazole-4-carbonitrile
Formel: C₅H₄N₂S
Molecular Weight: 124.164 g/mol
Schmelzpunkt: 58…61 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00068103
CAS-Nummer: 21917-76-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR