3-(2-Oxobenzooxazol-3-yl)propionic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(2-Oxo-2,3-dihydro-1,3-benzoxazol-3-yl)propanoic acid

OR29669-1G
APOSOR29669-1GEA 0 EUR
APOSOR29669-1G
3-(2-Oxobenzooxazol-3-yl)propionic acid
3-(2-Oxobenzooxazol-3-yl)propionic acid
Formel: C₁₀H₉NO₄
Molecular Weight: 207,19 g/mol
Schmelzpunkt: 125…126 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052166
CAS-Nummer: 13610-59-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR