Methyl-4-brom-1-ethyl-3-methyl-1H-pyrazol-5-carboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 4-bromo-1-ethyl-3-methyl-1H-pyrazole-5-carboxylate

OR29670-250MG
APOSOR29670-250MGEA 81.2 EUR
APOSOR29670-250MG
Methyl-4-brom-1-ethyl-3-methyl-1H-pyrazol-5-carboxylat
Methyl-4-brom-1-ethyl-3-methyl-1H-pyrazol-5-carboxylat
Formel: C₈H₁₁BrN₂O₂
Molecular Weight: 247,0891 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173775
CAS-Nummer: 175276-98-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR