Isoxazole-5-carboxamide
Lieferant: Apollo Scientific
|
Achtung
|
OR29674-1G
APOSOR29674-1GEA
348
EUR
APOSOR29674-1G
Isoxazole-5-carboxamide
Isoxazole-5-carboxamide
Formel:
C₄H₄N₂O₂ Schmelzpunkt: 173…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00276986 CAS-Nummer: 89032-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...