1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(tert-Butyl)-5- carboxy-3-methyl-1H-pyrazole

OR29682-1G
APOSOR29682-1GEA 0 EUR
APOSOR29682-1G
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
Formel: C₉H₁₄N₂O₂
Schmelzpunkt: 149…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00084917
CAS-Nummer: 175277-09-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR