1-tert-Butyl-3-methyl-1H-pyrazole-5-carbonyl chloride
Lieferant: Apollo Scientific
|
Gefahr
|
OR29683-250MG
APOSOR29683-250MGEA
39.3
EUR
APOSOR29683-250MG
1-tert-Butyl-3-methyl-1H-pyrazole-5-carbonyl chloride
1-tert-Butyl-3-methyl-1H-pyrazole-5-carbonyl chloride
Formel:
C₉H₁₃ClN₂O Molecular Weight: 200,665 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084915 CAS-Nummer: 175277-10-8 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...