Ethyl-5-(tert-butyl)isoxazole-3-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 5-(tert-butyl)isoxazole-3-carboxylate

OR29684-250MG
APOSOR29684-250MGEA 24.8 EUR
APOSOR29684-250MG
Ethyl-5-(tert-butyl)isoxazole-3-carboxylate
Ethyl-5-(tert-butyl)isoxazole-3-carboxylate
Formel: C₁₀H₁₅NO₃
Molecular Weight: 197,231 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00085057
CAS-Nummer: 91252-54-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR