3-(Hydroxymethyl)-5-(methylthio)-1,3,4-thiadiazole-2(3H)-thione

Lieferant: Apollo Scientific

OR29690-1G
APOSOR29690-1GEA 125 EUR
APOSOR29690-1G
3-(Hydroxymethyl)-5-(methylthio)-1,3,4-thiadiazole-2(3H)-thione
3-(Hydroxymethyl)-5-(methylthio)-1,3,4-thiadiazole-2(3H)-thione
Formel: C₄H₆N₂OS₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01027317
CAS-Nummer: 90567-39-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR