2-Methylthiazole-4-carboxamidine hydrochloride
Lieferant: Apollo Scientific
|
Achtung
|
OR29699-1G
APOSOR29699-1GEA
132
EUR
APOSOR29699-1G
2-Methylthiazole-4-carboxamidine hydrochloride
2-Methylthiazole-4-carboxamidine hydrochloride
Formel:
C₅H₈ClN₃S Molecular Weight: 177,66 g/mol Schmelzpunkt: 185…189 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00178765 CAS-Nummer: 18876-82-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...