4-(Morpholino)phenylboronsäurepinakolester
Lieferant: Apollo Scientific
|
Achtung
|
OR2972-250MG
APOSOR2972-250MGEA
0
EUR
APOSOR2972-250MG
4-(Morpholino)phenylboronsäurepinakolester
4-(Morpholino)phenylboronsäurepinakolester
Formel:
C₁₆H₂₄BNO₃ Schmelzpunkt: 93…95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04112544 CAS-Nummer: 568577-88-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...