Isoxazole-5-thiocarboxamide

Lieferant: Apollo Scientific
Achtung

Synonyme: Isoxazole-5-carbothioic acid amide

OR29726-250MG
APOSOR29726-250MGEA 174 EUR
APOSOR29726-250MG
Isoxazole-5-thiocarboxamide
Isoxazole-5-thiocarboxamide
Formel: C₄H₄N₂OS
Molecular Weight: 128,152 g/mol
Schmelzpunkt: 160…164 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00276995
CAS-Nummer: 175334-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR