N-(3,5-Dichlorphenyl)-2-chloracetamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(Chloroacetyl)-3,5-dichloroaniline
OR29761-1G
APOSOR29761-1GEA
84.1
EUR
APOSOR29761-1G
N-(3,5-Dichlorphenyl)-2-chloracetamid
N-(3,5-Dichlorphenyl)-2-chloracetamid
Formel:
C₈H₆Cl₃NO Schmelzpunkt: 140…142 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
33560-48-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...