4-Chlorbenzhydrazid 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chlorobenzoic hydrazide
OR29769-25G
APOSOR29769-25GEA
59.6
EUR
APOSOR29769-25G
4-Chlorbenzhydrazid 97%
4-Chlorbenzhydrazid
Formel:
C₇H₇ClN₂O Molecular Weight: 170,6 g/mol Schmelzpunkt: 162…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007603 CAS-Nummer: 536-40-3 EINECS: 208-632-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...