Ethyl-2-chlorisonicotinat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-Chloropyridine-4-carboxylate

OR2977-100G
APOSOR2977-100GEA 136 EUR
APOSOR2977-100G
Ethyl-2-chlorisonicotinat
Ethyl-2-chlorisonicotinat
Formel: C₈H₈ClNO₂
Molecular Weight: 185.6076 g/mol
Siedepunkt: 130…132 °C/20mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD04114259
CAS-Nummer: 54453-93-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR