2-Cyanothioacetamide
Lieferant: Apollo Scientific
|
Achtung
|
OR29773-100G
OR29773-25G
OR29773-5G
APOSOR29773-100GEA
157
EUR
APOSOR29773-100G
APOSOR29773-25G
APOSOR29773-5G
2-Cyanothioacetamide
2-Cyanothioacetamide
Formel:
C₃H₄N₂S Schmelzpunkt: 114…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010025 CAS-Nummer: 7357-70-2 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...