2-(4-tert-Butylphenoxy)ethanimidamide hydrochloride

Lieferant: Apollo Scientific

OR29774-1G
APOSOR29774-1GEA 109 EUR
APOSOR29774-1G
2-(4-tert-Butylphenoxy)ethanimidamide hydrochloride
2-(4-tert-Butylphenoxy)ethanimidamide hydrochloride
Formel: C₁₂H₁₉ClN₂O
Molecular Weight: 242,75 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173807
CAS-Nummer: 175277-47-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR