6-(4-Chlorophenoxy)nicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR29796-100MG
APOSOR29796-100MGEA 21.9 EUR
APOSOR29796-100MG
6-(4-Chlorophenoxy)nicotinonitrile
6-(4-Chlorophenoxy)nicotinonitrile
Formel: C₁₂H₇ClN₂O
Molecular Weight: 230,65 g/mol
Schmelzpunkt: 129…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00221203
CAS-Nummer: 99902-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR