2-(Phenylthio)ethanimidamide hydrochloride

Lieferant: Apollo Scientific
Achtung

OR29804-1G
APOSOR29804-1GEA 98.6 EUR
APOSOR29804-1G
2-(Phenylthio)ethanimidamide hydrochloride
2-(Phenylthio)ethanimidamide hydrochloride
Formel: C₈H₁₁ClN₂S
Schmelzpunkt: 175…177 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00100031
CAS-Nummer: 84544-86-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR