4-Chlorbenzamidin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Chlorbenzoesäureamidin Hydrochlorid, 4-Chlorphenylamidin Hydrochlorid

OR29823-100G
APOSOR29823-100GEA 207 EUR
APOSOR29823-100G
4-Chlorbenzamidin Hydrochlorid
4-Chlorbenzamidin Hydrochlorid
Formel: C₇H₈Cl₂N₂
Molecular Weight: 191.06 g/mol
Schmelzpunkt: 244…245 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00126401
CAS-Nummer: 14401-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR