2-Chlorisonicotinsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chlorpyridin-4-carbonsäure, 2-Chlor-4-pyridincarbonsäure

OR29861-100G OR29861-1G OR29861-5G
APOSOR29861-100GEA 125 EUR
APOSOR29861-100G APOSOR29861-1G APOSOR29861-5G
2-Chlorisonicotinsäure
2-Chlorisonicotinsäure
Formel: C₆H₄ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00191402
CAS-Nummer: 6313-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR