5-Chlor-2-hydroxynicotinsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloro-2-hydroxy-3-pyridinecarboxylic Acid
OR29878-25G
APOSOR29878-25GEA
63.8
EUR
APOSOR29878-25G
5-Chlor-2-hydroxynicotinsäure 97%
5-Chlor-2-hydroxynicotinsäure
Formel:
C₆H₄ClNO₃ Schmelzpunkt: 258…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00204158 CAS-Nummer: 38076-80-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...